MAT047021447 Matrix Scientific

5-Fluoro-4-hydroxyquinazoline

  • MDL number MFCD00173674
  • Cas Number : 436-72-6
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-242
Country of Origin USA
Product Link http://www.matrixscientific.com/002541.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002541.html
Availability details for
Quantity
  • Availability
  • Add to Cart