MAT047021456 Matrix Scientific

3-Chloro-4-iodonitrobenzene

  • MDL number MFCD00024589
  • Cas Number : 41252-96-4
  • Molecular Weight : 283.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/002550.html

Safety Information

hcodes Irritant/Light Sensitive
MSDS http://www.matrixscientific.com/002550.html
Availability details for
Quantity
  • Availability
  • Add to Cart