MAT047021466 Matrix Scientific

3-Chloro-4-methoxytoluene

  • MDL number MFCD00060649
  • Cas Number : 22002-44-4
  • Molecular Weight : 156.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 212-217/748 Torr
Product Link http://www.matrixscientific.com/002563.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002563.html
Availability details for
Quantity
  • Availability
  • Add to Cart