MAT047021473 Matrix Scientific

3-Fluoro-4-methylbenzamide oxime

  • MDL number MFCD00673178
  • Cas Number : 238742-80-8
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Country of Origin USA
Product Link http://www.matrixscientific.com/002571.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002571.html
Availability details for
Quantity
  • Availability
  • Add to Cart