MAT047021476 Matrix Scientific

3-Fluoro-4-methylbenzamide

  • MDL number MFCD01631412
  • Cas Number : 170726-98-4
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002574.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002574.html
Availability details for
Quantity
  • Availability
  • Add to Cart