MAT047021477 Matrix Scientific

3-Fluoro-4-methylbenzoyl chloride

  • MDL number MFCD00671774
  • Cas Number : 59189-97-8
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002575.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/002575.html
Availability details for
Quantity
  • Availability
  • Add to Cart