MAT047021479 Matrix Scientific

3-Fluoro-4-methylbenzylamine

  • MDL number MFCD01631531
  • Cas Number : 261951-67-1
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002577.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/002577.html
Availability details for
Quantity
  • Availability
  • Add to Cart