MAT047021482 Matrix Scientific

3-Fluoro-4-methylcinnamic acid

  • MDL number MFCD01631413
  • Cas Number : 261951-72-8
  • Molecular Weight : 180.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002580.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002580.html
Availability details for
Quantity
  • Availability
  • Add to Cart