MAT047021484 Matrix Scientific

3-Fluoro-4-methylphenylacetonitrile

  • MDL number MFCD01631535
  • Cas Number : 261951-73-9
  • Molecular Weight : 149.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002582.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002582.html
Availability details for
Quantity
  • Availability
  • Add to Cart