MAT047021486 Matrix Scientific

3-Fluoro-4-methylphenylacetic acid

  • MDL number MFCD01631536
  • Cas Number : 261951-74-0
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002583.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002583.html
Availability details for
Quantity
  • Availability
  • Add to Cart