MAT047021494 Matrix Scientific

2-Chloro-4-nitrobenzhydrazide

  • MDL number MFCD00017047
  • Cas Number : 67345-78-2
  • Molecular Weight : 215.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Storage temp. Argon
Country of Origin USA
Product Link http://www.matrixscientific.com/002592.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/002592.html
Availability details for
Quantity
  • Availability
  • Add to Cart