MAT047021516 Matrix Scientific

2-Fluoro-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD00061280
  • Cas Number : 4088-84-0
  • Molecular Weight : 189.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 185-187
Product Link http://www.matrixscientific.com/002615.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002615.html
Availability details for
Quantity
  • Availability
  • Add to Cart