MAT047021527 Matrix Scientific

3-Fluoro-5-(trifluoromethyl)benzophenone

  • MDL number MFCD00061256
  • Cas Number : 239087-02-6
  • Molecular Weight : 268.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002627.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002627.html
Availability details for
Quantity
  • Availability
  • Add to Cart