MAT047021530 Matrix Scientific

1,1,1,2,3,3-Hexafluoropropyl methyl ether

  • MDL number MFCD00041537
  • Cas Number : 382-34-3
  • Molecular Weight : 182.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.419
Country of Origin USA
Boiling point 53°
Product Link http://www.matrixscientific.com/001191.html

Safety Information

MSDS http://www.matrixscientific.com/001191.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart