MAT047021533 Matrix Scientific

2-Fluoro-5-(trifluoromethyl)benzyl bromide

  • MDL number MFCD00061174
  • Cas Number : 220239-69-0
  • Molecular Weight : 257.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/002632.html

Safety Information

hcodes Corrosive/Lachrymator
MSDS http://www.matrixscientific.com/002632.html
Availability details for
Quantity
  • Availability
  • Add to Cart