MAT047021554 Matrix Scientific

2-Chloro-5-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD01631471
  • Cas Number : 22893-39-6
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/002651.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002651.html
Availability details for
Quantity
  • Availability
  • Add to Cart