MAT047021566 Matrix Scientific

Methyl 2,5-bis(trifluoroethoxy)benzoate

  • MDL number MFCD00236693
  • Cas Number : 35480-31-0
  • Molecular Weight : 332.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/002679.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002679.html
Availability details for
Quantity
  • Availability
  • Add to Cart