MAT047021571 Matrix Scientific

Dimethyl 5-chloroisophthalate

  • MDL number MFCD00079752
  • Cas Number : 20330-90-9
  • Molecular Weight : 228.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/002684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002684.html
Availability details for
Quantity
  • Availability
  • Add to Cart