MAT047021577 Matrix Scientific

2-Chloro-5-fluoroaniline

  • MDL number MFCD00084852
  • Cas Number : 452-83-5
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26
Purity 97
Country of Origin USA
Boiling point 211
Product Link http://www.matrixscientific.com/002689.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002689.html
Availability details for
Quantity
  • Availability
  • Add to Cart