MAT047021584 Matrix Scientific

2-Chloro-5-fluorobenzyl alcohol

  • MDL number MFCD01631418
  • Cas Number : 261762-59-8
  • Molecular Weight : 160.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-72
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002698.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002698.html
Availability details for
Quantity
  • Availability
  • Add to Cart