MAT047021590 Matrix Scientific

Dimethyl 5-iodoisophthalate

  • MDL number MFCD00079753
  • Cas Number : 51839-15-7
  • Molecular Weight : 320.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-104
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/002705.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002705.html
Availability details for
Quantity
  • Availability
  • Add to Cart