MAT047021602 Matrix Scientific

2-Methyl-5-nitrobenzotrifluoride

  • MDL number MFCD01631684
  • Cas Number : 89976-12-5
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Country of Origin USA
Product Link http://www.matrixscientific.com/002719.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002719.html
Availability details for
Quantity
  • Availability
  • Add to Cart