MAT047021618 Matrix Scientific

Silver(I) fluoride

  • MDL number MFCD00003410
  • Cas Number : 7775-41-9
  • Molecular Weight : 126.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 435
Purity 99
Density 5.852
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 1159
Product Link http://www.matrixscientific.com/001028.html

Safety Information

hcodes Corrosive/Stored under Argon/Light Sensitive/Hygroscopic
MSDS http://www.matrixscientific.com/001028.html
Availability details for
Quantity
  • Availability
  • Add to Cart