MAT047021645 Matrix Scientific

2-Chloro-6-fluoro-3-methylphenylacetonitrile

  • MDL number MFCD01631364
  • Cas Number : 261762-94-1
  • Molecular Weight : 183.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002763.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002763.html
Availability details for
Quantity
  • Availability
  • Add to Cart