MAT047021649 Matrix Scientific

2-Chloro-6-fluorobenzoyl chloride

  • MDL number MFCD00178746
  • Cas Number : 79455-63-3
  • Molecular Weight : 193.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002768.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/002768.html
Availability details for
Quantity
  • Availability
  • Add to Cart