MAT047021668 Matrix Scientific

1-Chloro-6-iodoperfluorohexane

  • MDL number MFCD01320787
  • Cas Number : 16486-97-8
  • Molecular Weight : 462.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 68� Torr
Product Link http://www.matrixscientific.com/002787.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002787.html
Availability details for
Quantity
  • Availability
  • Add to Cart