MAT047021675 Matrix Scientific

2-Fluoro-6-phenoxybenzonitrile

  • MDL number MFCD00068201
  • Cas Number : 175204-06-5
  • Molecular Weight : 213.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-93
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/002798.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002798.html
Availability details for
Quantity
  • Availability
  • Add to Cart