MAT047021703 Matrix Scientific

1H,1H,9H-Hexadecafluoro-1-nonanol

  • MDL number MFCD00039629
  • Cas Number : 376-18-1
  • Molecular Weight : 432.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66
Purity 97
Country of Origin USA
Boiling point 157-158�0mm
Product Link http://www.matrixscientific.com/002834.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002834.html
Availability details for
Quantity
  • Availability
  • Add to Cart