MAT047021708 Matrix Scientific

Thallium(I) fluoride

  • MDL number MFCD00049607
  • Cas Number : 7789-27-7
  • Molecular Weight : 223.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 327
Purity 99
Density 8.36/20
Country of Origin USA
Boiling point 665
Product Link http://www.matrixscientific.com/002838.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002838.html
Availability details for
Quantity
  • Availability
  • Add to Cart