MAT047021721 Matrix Scientific

N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine

  • MDL number MFCD00052691
  • Cas Number : 120940-43-4
  • Molecular Weight : 235.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Country of Origin USA
Product Link http://www.matrixscientific.com/002853.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/002853.html
Availability details for
Quantity
  • Availability
  • Add to Cart