MAT047021733 Matrix Scientific

1H,1H,2H-Perfluoro-1-dodecene

  • MDL number MFCD00042346
  • Cas Number : 30389-25-4
  • Molecular Weight : 546.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Density 1.711
Country of Origin USA
Boiling point 71-72�mm
Product Link http://www.matrixscientific.com/002864.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/002864.html
Availability details for
Quantity
  • Availability
  • Add to Cart