MAT047021743 Matrix Scientific

3,4,5-Trifluoroacetophenone

  • MDL number MFCD00083533
  • Cas Number : 220141-73-1
  • Molecular Weight : 174.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point >100
Country of Origin USA
Product Link http://www.matrixscientific.com/002876.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002876.html
Availability details for
Quantity
  • Availability
  • Add to Cart