MAT047021756 Matrix Scientific

2',4',6'-Trifluoropropiophenone

  • MDL number MFCD00061224
  • Cas Number : 220141-69-5
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 67-69�m
Product Link http://www.matrixscientific.com/002889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002889.html
Availability details for
Quantity
  • Availability
  • Add to Cart