MAT047021810 Matrix Scientific

Tris(heptafluoropropyl)-s-triazine

  • MDL number MFCD00042438
  • Cas Number : 915-76-4
  • Molecular Weight : 585.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35
Density 1.716/25
Country of Origin USA
Boiling point 165
Product Link http://www.matrixscientific.com/002954.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002954.html
Availability details for
Quantity
  • Availability
  • Add to Cart