MAT047021823 Matrix Scientific

4-Fluoromandelic acid

  • MDL number MFCD00044402
  • Cas Number : 395-33-5
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/002973.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002973.html
Availability details for
Quantity
  • Availability
  • Add to Cart