MAT047021832 Matrix Scientific

Ethyl beta-naphthoate

  • MDL number MFCD00016811
  • Cas Number : 3007-91-8
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32
Purity 99
Country of Origin USA
Boiling point 308-309
Product Link http://www.matrixscientific.com/002985.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002985.html
Availability details for
Quantity
  • Availability
  • Add to Cart