MAT047021858 Matrix Scientific

7-Methoxybenzofuran-2-carboxylic acidethyl ester

  • MDL number MFCD00060510
  • Cas Number : 50551-58-1
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-84
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/003016.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003016.html
Availability details for
Quantity
  • Availability
  • Add to Cart