MAT047021895 Matrix Scientific

2,2,3,3,4,4-Hexafluoro-1,5-pentyl diacrylate

  • MDL number MFCD02093323
  • Cas Number : 678-95-5
  • Molecular Weight : 320.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/001229.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/001229.html
Availability details for
Quantity
  • Availability
  • Add to Cart