MAT047021905 Matrix Scientific

Methyl 4-chloro-2,2-dimethyl-4-pentenoate

  • MDL number MFCD00075437
  • Cas Number : 86799-85-1
  • Molecular Weight : 176.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.05
Country of Origin US
Boiling point 97-98�mm
Product Link https://www.matrixscientific.com/003081.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart