MAT047021915 Matrix Scientific

Methyl 4-chloro-6-(trifluoromethyl)thieno[3,4-b]-pyridine-1-carboxylate

  • MDL number MFCD00173840
  • Cas Number : 175203-40-4
  • Molecular Weight : 295.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-106
Country of Origin USA
Product Link http://www.matrixscientific.com/003090.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003090.html
Availability details for
Quantity
  • Availability
  • Add to Cart