MAT047021918 Matrix Scientific

2,4,6-Trichlorobenzonitrile

  • MDL number MFCD00052711
  • Cas Number : 6575-05-9
  • Molecular Weight : 206.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/003092.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/003092.html
Availability details for
Quantity
  • Availability
  • Add to Cart