MAT047021933 Matrix Scientific

Malonic acid dihydrazide

  • MDL number MFCD00041268
  • Cas Number : 3815-86-9
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/003109.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003109.html
Availability details for
Quantity
  • Availability
  • Add to Cart