MAT047021972 Matrix Scientific

4-Acetamidophenylacetonitrile

  • MDL number MFCD00016398
  • Cas Number : 25025-06-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Country of Origin USA
Product Link http://www.matrixscientific.com/003156.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003156.html
Availability details for
Quantity
  • Availability
  • Add to Cart