MAT047022005 Matrix Scientific

3-(Cyclopentylamino)propionitrile

  • MDL number MFCD00060286
  • Cas Number : 1074-63-1
  • Molecular Weight : 138.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 0.95
Country of Origin USA
Boiling point 115-116�m
Product Link http://www.matrixscientific.com/003198.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003198.html
Availability details for
Quantity
  • Availability
  • Add to Cart