MAT047022028 Matrix Scientific

5-Bromo-2,4-difluorobenzenesulfonyl chloride

  • MDL number MFCD01940424
  • Cas Number : 287172-61-6
  • Molecular Weight : 291.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-36
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/001243.html

Safety Information

hcodes Corrosive/Moisture Sensitive
MSDS http://www.matrixscientific.com/001243.html
Availability details for
Quantity
  • Availability
  • Add to Cart