MAT047022032 Matrix Scientific

3-Aminophenyl trifluoromethyl sulfide

  • MDL number MFCD00190130
  • Cas Number : 369-68-6
  • Molecular Weight : 193.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.342
Country of Origin USA
Boiling point 221
Product Link http://www.matrixscientific.com/003231.html

Safety Information

hcodes Toxic/Stench
MSDS http://www.matrixscientific.com/003231.html
Availability details for
Quantity
  • Availability
  • Add to Cart