MAT047022043 Matrix Scientific

4-Nitrophenyloxamic acid

  • MDL number MFCD00014709
  • Cas Number : 103-94-6
  • Molecular Weight : 210.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210�ec)
Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/003247.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003247.html
Availability details for
Quantity
  • Availability
  • Add to Cart