MAT047022054 Matrix Scientific

4-Fluorobenzamide

  • MDL number MFCD00007992
  • Cas Number : 824-75-9
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-156
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/003263.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003263.html
Availability details for
Quantity
  • Availability
  • Add to Cart