MAT047022075 Matrix Scientific

5-Fluorobenzo-[2,1,3]-thiadiazole

  • MDL number MFCD00173946
  • Cas Number : 17821-75-9
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 96�mm
Product Link http://www.matrixscientific.com/003286.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003286.html
Availability details for
Quantity
  • Availability
  • Add to Cart