MAT047022088 Matrix Scientific

2-Fluorobenzophenone

  • MDL number MFCD00000318
  • Cas Number : 342-24-5
  • Molecular Weight : 200.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.186
Country of Origin USA
Boiling point 150� Torr
Product Link http://www.matrixscientific.com/003301.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/003301.html
Availability details for
Quantity
  • Availability
  • Add to Cart